Gene Transcripts

Gene Symbol
Mbd2
Entrez Gene ID
17191
Gene Name
methyl-CpG binding domain protein 2
Gene Aliases
MBD2a
Chromosome Location
Chr. 18: 70568270 - 70626131 on Build GRCm38
UniGene ID
Mm.322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010773.2 NP_034903.2 3 933
GenBank mRNA AF072243.1 - 3 891
AK035891.1 - 1 1412
AK147008.1 - 3 818
BC026698.1 - 3 893
BC046607.1 - 3 933 6399206
BC057289.1 - 3 765
BC058864.1 - 3 753

View Full Product Details