Gene Transcripts

Gene Symbol
Mcpt8
Entrez Gene ID
17231
Gene Name
mast cell protease 8
Gene Aliases
BB139338, MMCP-8
Chromosome Location
Chr. 14: 56082163 - 56085276 on Build GRCm38
UniGene ID
Mm.41979
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008572.1 NP_032598.1 3 241
GenBank mRNA AK136485.1 - 3 261
BC119342.1 - 3 317 8734091
BC120607.1 - 3 314 40129530
BC144934.1 - 3 403
X78545.1 - 3 241

View Full Product Details