Gene Transcripts

Gene Symbol
Mdm1
Entrez Gene ID
17245
Gene Name
transformed mouse 3T3 cell double minute 1
Gene Aliases
Arrd2, Mdm-1
Chromosome Location
Chr. 10: 118141537 - 118168999 on Build GRCm38
UniGene ID
Mm.101191
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162904.1 NP_001156376.1 2 278
NM_001162905.1 NP_001156377.1 2 278
NM_010785.2 NP_034915.2 2 278
NM_148922.3 NP_683724.2 2 278
GenBank mRNA AK004789.2 - 2 233
BC052680.1 - 2 228
BC137608.1 - 2 206 8860627
BC144777.1 - 2 206 9053325
EU490427.1 - 2 120
EU490428.1 - 2 120
M20823.1 - 2 141
M20824.1 - 2 141

View Full Product Details