Gene Transcripts

Gene Symbol
Meox1
Entrez Gene ID
17285
Gene Name
mesenchyme homeobox 1
Gene Aliases
AI385561, D330041M02Rik, Mox-1, Mox1
Chromosome Location
Chr. 11: 101877510 - 101894354 on Build GRCm38
UniGene ID
Mm.3404
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010791.3 NP_034921.1 2 962
GenBank mRNA AK052370.1 - 2 972
AK133672.1 - 2 333
BC011082.1 - 2 962 3984366
Z15103.1 - 2 917

View Full Product Details