Gene Transcripts

Gene Symbol
Marcksl1
Entrez Gene ID
17357
Gene Name
MARCKS-like 1
Gene Aliases
AL022768, AW215397, AW536807, D4Bc1, F52, MacMARCKS, Macs2, Macs3, Mlp, Mrp
Chromosome Location
Chr. 4: 129513581 - 129515981 on Build GRCm38
UniGene ID
Mm.424974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010807.4 NP_034937.1 2 562
GenBank mRNA AK079390.1 - 2 511
AK083913.1 - 2 513
AK150261.1 - 2 509
AK152990.1 - 2 510
AK169355.1 - 2 513
BC006757.1 - 2 490 3257582
X61399.1 - 2 499

View Full Product Details