Gene Transcripts

Gene Symbol
Msx3
Entrez Gene ID
17703
Gene Name
msh homeobox 3
Gene Aliases
AI323377
Chromosome Location
Chr. 7: 140046156 - 140049970 on Build GRCm38
UniGene ID
Mm.4816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010836.3 NP_034966.1 2 462
GenBank mRNA AB221640.1 - 2 373
AK012208.1 - 2 461
AK012556.1 - 2 531
BC051983.1 - 2 431
BC137577.1 - 2 461 8860596
BC137578.1 - 2 461 8860597
U62523.1 - 2 469

View Full Product Details