Gene Transcripts

Gene Symbol
HTR1B
Entrez Gene ID
3351
Gene Name
5-hydroxytryptamine receptor 1B
Gene Aliases
5-HT-1B, 5-HT-1D-beta, 5-HT1B, 5-HT1DB, HTR1D2, HTR1DB, S12
Chromosome Location
Chr. 6: 77460848 - 77464022 on Build GRCh38
UniGene ID
Hs.123016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000863.2 NP_000854.1 1 1743
GenBank mRNA AK290080.1 - 1 1494
AK315325.1 - 1 1494
BC069065.1 - 1 1124 7216878
BC096206.1 - 1 1124
BC096207.1 - 1 1124 40004711
BC096208.1 - 1 1124
KJ534870.1 - 1 1124
M81590.1 - 1 1185

View Full Product Details