Gene Transcripts
- Gene Symbol
- Ndp
- Entrez Gene ID
- 17986
- Gene Name
- Norrie disease (pseudoglioma) (human)
- Gene Aliases
- ND, Ndph
- Chromosome Location
- Chr. X: 16885521 - 16911764 on Build GRCm38
- UniGene ID
- Mm.5014
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC090623.1 |
- |
2 |
662 |
|
X92397.1 |
- |
2 |
508 |
|
View Full Product Details