Gene Transcripts

Gene Symbol
Nek2
Entrez Gene ID
18005
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 2
Gene Aliases
AA617254, C77054
Chromosome Location
Chr. 1: 191820875 - 191833049 on Build GRCm38
UniGene ID
Mm.33773
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010892.3 NP_035022.2 3 662
GenBank mRNA AF007247.1 - 3 576
AF013166.1 - 3 546
AK077627.1 - 3 626
AK147072.1 - 3 638
AK164467.1 - 3 605
AK169330.1 - 4 553
AK170179.1 - 3 582
AK171277.1 - 3 582
AK171387.1 - 3 582
BC010302.1 - 3 584 3597227
BC011316.1 - 3 582 3674001
BC057576.1 - 3 602 6810914
U95610.1 - 3 573

View Full Product Details