Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008698.2 | NP_032724.1 | 5 | 419 | |
GenBank mRNA | AJ001260.1 | - | 5 | 419 | |
AK013140.1 | - | 5 | 427 | ||
AK031804.1 | - | 5 | 435 | ||
AK147786.1 | - | 6 | 696 | ||
BC010837.1 | - | 5 | 417 | 4242309 |