Gene Transcripts
- Gene Symbol
- Odf2
- Entrez Gene ID
- 18286
- Gene Name
- outer dense fiber of sperm tails 2
- Gene Aliases
- AI848335, MMTEST29
- Chromosome Location
- Chr. 2: 29889720 - 29931746 on Build GRCm38
- UniGene ID
- Mm.330116
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113213.1 |
NP_001106684.1
|
13, 14 |
1735 |
|
NM_001113214.1 |
NP_001106685.1
|
13, 14 |
1735 |
|
NM_001177659.1 |
NP_001171130.1
|
13, 14 |
1738 |
|
NM_001177661.1 |
NP_001171132.1
|
12, 13 |
1581 |
|
NM_013615.3 |
NP_038643.1
|
13, 14 |
1708 |
|
GenBank mRNA |
AF000968.1 |
- |
11, 12 |
1555 |
|
AF034105.1 |
- |
13, 14 |
1671 |
|
AK143726.1 |
- |
13, 14 |
1642 |
|
AK148523.1 |
- |
13, 14 |
1648 |
|
AK168415.1 |
- |
13, 14 |
1637 |
|
BC057001.1 |
- |
14, 15 |
1792 |
6827260 |
BC067406.1 |
- |
3 |
2717 |
|
View Full Product Details