Gene Transcripts
- Gene Symbol
- Olfr8
- Entrez Gene ID
- 18372
- Gene Name
- olfactory receptor 8
- Gene Aliases
- GA_x5J8B7TNKDU-1250-1574, MOR139-5P
- Chromosome Location
- Chr. 10: 78955207 - 78956139 on Build GRCm38
- UniGene ID
- Mm.391567
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207201.1 |
NP_997084.1
|
1 |
509 |
|
- Gene Symbol
- Olfr1354
- Entrez Gene ID
- 259163
- Gene Name
- olfactory receptor 1354
- Gene Aliases
- EG257869, GA_x5J8B7TPDN2-1058-1265, GA_x6K02T03FR9-4826-3919, MOR139-5, MOR139-7, MOR185-8, Olfr1354-ps1, Olfr233-ps1
- Chromosome Location
- Chr. 10: 78916842 - 78917936 on Build GRCm38
- UniGene ID
- Mm.419145
- Species
- Mus musculus
- Gene Symbol
- Olfr1355
- Entrez Gene ID
- 257734
- Gene Name
- olfactory receptor 1355
- Gene Aliases
- GA_x5J8B7W4DUT-12263-11330, MOR139-6
- Chromosome Location
- Chr. 10: 78875549 - 78880106 on Build GRCm38
- UniGene ID
- Mm.334149
- Species
- Mus musculus
View Full Product Details