Gene Transcripts

Gene Symbol
Olfr9
Entrez Gene ID
18373
Gene Name
olfactory receptor 9
Gene Aliases
MOR269-3
Chromosome Location
Chr. 10: 128989914 - 128990852 on Build GRCm38
UniGene ID
Mm.377111
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146861.1 NP_667072.1 1 298
GenBank mRNA BC116994.1 - 1 363
BC119365.1 - 1 363 8734114

Gene Symbol
Olfr763
Entrez Gene ID
258861
Gene Name
olfactory receptor 763
Gene Aliases
MOR269-2
Chromosome Location
Chr. 10: 129011287 - 129012216 on Build GRCm38
UniGene ID
Mm.377724
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146862.1 NP_667073.1 1 298
GenBank mRNA BC119229.1 - 1 363
BC119231.1 - 1 363 8733980

View Full Product Details