Gene Transcripts

Gene Symbol
Otc
Entrez Gene ID
18416
Gene Name
ornithine transcarbamylase
Gene Aliases
AI265390, Sf, spf
Chromosome Location
Chr. X: 10252297 - 10321024 on Build GRCm38
UniGene ID
Mm.2611
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008769.3 NP_032795.1 4, 5 468
GenBank mRNA AK050930.1 - 4, 5 469
BC024893.1 - 4, 5 460 4196455
M17030.1 - 4, 5 394

View Full Product Details