Gene Transcripts

Gene Symbol
Pax7
Entrez Gene ID
18509
Gene Name
paired box 7
Gene Aliases
Pax-7
Chromosome Location
Chr. 4: 139737059 - 139833026 on Build GRCm38
UniGene ID
Mm.218760
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011039.2 NP_035169.1 2 316
GenBank mRNA AF254422.4 - 2 316
U20792.1 - 1 157

View Full Product Details