Gene Transcripts

Gene Symbol
Rbpjl
Entrez Gene ID
19668
Gene Name
recombination signal binding protein for immunoglobulin kappa J region-like
Gene Aliases
RBP-L, Rbpsuhl
Chromosome Location
Chr. 2: 164403056 - 164415448 on Build GRCm38
UniGene ID
Mm.473794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009036.1 NP_033062.1 8, 9 965
GenBank mRNA AK052449.1 - 6, 7 4076
AK131814.1 - 8, 9 946
BC152311.1 - 8, 9 959 40090714
Y10926.1 - 8, 9 965

View Full Product Details