Gene Transcripts

Gene Symbol
Bex1
Entrez Gene ID
19716
Gene Name
brain expressed gene 1
Gene Aliases
AA407839, AU018692, Bex4, Rex3
Chromosome Location
Chr. X: 136213972 - 136215513 on Build GRCm38
UniGene ID
Mm.422943
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009052.2 NP_033078.2 3 824
GenBank mRNA AF051347.1 - 3 757
AF097438.1 - 3 769
AK003429.1 - 1 439
BC058805.1 - 1 1096
BC061179.1 - 3 769 6531808
BC089464.1 - 3 818 6594252
CB194115.1 - 3 746

View Full Product Details