Gene Transcripts
- Gene Symbol
- Sord
- Entrez Gene ID
- 20322
- Gene Name
- sorbitol dehydrogenase
- Gene Aliases
- Sdh-1, Sdh1, Sodh-1
- Chromosome Location
- Chr. 2: 122234839 - 122265337 on Build GRCm38
- UniGene ID
- Mm.371580
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146126.4 |
NP_666238.1
|
1, 2 |
130 |
|
GenBank mRNA |
AK004692.1 |
- |
1, 2 |
154 |
|
AK015059.1 |
- |
1, 2 |
131 |
|
AK152169.1 |
- |
1, 2 |
179 |
|
AK166988.1 |
- |
1, 2 |
130 |
|
AK166996.1 |
- |
1, 2 |
159 |
|
AK209251.1 |
- |
1 |
16 |
|
BC024124.1 |
- |
1, 2 |
145 |
5099395 |
BC030875.1 |
- |
1, 2 |
119 |
4237164 |
BC039957.1 |
- |
1, 2 |
130 |
|
BC092291.1 |
- |
1, 2 |
133 |
5097505 |
U27014.1 |
- |
|
130 |
|
View Full Product Details