Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011348.2 | NP_035478.2 | 3 | 932 | |
GenBank mRNA | AF034744.1 | - | 3 | 667 | |
AK141295.1 | - | 3 | 934 | ||
AK142340.1 | - | 3 | 759 | ||
AK159214.1 | - | 3 | 723 | ||
BC057956.1 | - | 3 | 830 | 5357516 | |
Z80941.1 | - | 3 | 298 | ||
Z93947.1 | - | 3 | 907 | ||
Z93948.1 | - | 3 | 907 |