Gene Transcripts
- Gene Symbol
- IL12RB2
- Entrez Gene ID
- 3595
- Gene Name
- interleukin 12 receptor subunit beta 2
- Gene Aliases
- Chromosome Location
- Chr. 1: 67307351 - 67397730 on Build GRCh38
- UniGene ID
- Hs.479347
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001258214.1 |
NP_001245143.1
|
5, 6 |
1202 |
|
NM_001258215.1 |
NP_001245144.1
|
4, 5 |
1114 |
|
NM_001258216.1 |
NP_001245145.1
|
4, 5 |
1114 |
|
NM_001319233.1 |
NP_001306162.1
|
5, 6 |
1198 |
|
NM_001559.2 |
NP_001550.1
|
4, 5 |
1114 |
|
NR_047583.1 |
|
4, 5 |
1114 |
|
NR_047584.1 |
|
4, 5 |
1114 |
|
XM_005270825.2 |
XP_005270882.1
|
5, 6 |
1215 |
|
XM_005270827.2 |
XP_005270884.1
|
5, 6 |
1211 |
|
XM_005270828.3 |
XP_005270885.1
|
4, 5 |
1123 |
|
XM_006710617.2 |
XP_006710680.1
|
5, 6 |
1215 |
|
XM_011541383.2 |
XP_011539685.1
|
5, 6 |
667 |
|
XM_011541384.2 |
XP_011539686.1
|
5, 6 |
1215 |
|
XM_017001202.1 |
XP_016856691.1
|
5, 6 |
1215 |
|
XM_017001203.1 |
XP_016856692.1
|
4, 5 |
1127 |
|
XM_017001204.1 |
XP_016856693.1
|
5, 6 |
1215 |
|
GenBank mRNA |
AK316525.1 |
- |
5, 6 |
673 |
|
BC104772.1 |
- |
3, 4 |
524 |
8143775 |
BC104774.1 |
- |
3, 4 |
524 |
8143777 |
BC143248.1 |
- |
3, 4 |
524 |
|
BC143249.1 |
- |
3, 4 |
524 |
9051751 |
BC143250.1 |
- |
3, 4 |
524 |
|
U64198.1 |
- |
4, 5 |
1114 |
|
View Full Product Details