Gene Transcripts

Gene Symbol
Ostf1
Entrez Gene ID
20409
Gene Name
osteoclast stimulating factor 1
Gene Aliases
C78236, SH3P2, Sh3d3
Chromosome Location
Chr. 19: 18580364 - 18631813 on Build GRCm38
UniGene ID
Mm.172222
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017375.3 NP_059071.1 9 724
GenBank mRNA AK002899.1 - 9 703
AK005525.1 - 9 693
AK008123.1 - 9 672
AK008467.1 - 9 672
AK010074.1 - 9 720
AK145949.1 - 9 675
AK149175.1 - 702
AK150145.1 - 9 694
AK155028.1 - 9 694
BC060986.1 - 9 603 30300544
U58888.1 - 9 672

View Full Product Details