Gene Transcripts

Gene Symbol
Shcbp1
Entrez Gene ID
20419
Gene Name
Shc SH2-domain binding protein 1
Gene Aliases
mPAL
Chromosome Location
Chr. 8: 4735980 - 4779534 on Build GRCm38
UniGene ID
Mm.37801
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011369.2 NP_035499.1 9 1296
GenBank mRNA AF017152.1 - 9 1296
AK135226.1 - 1 1918
AK137496.1 - 8 1778
AK144966.1 - 9 1319
AK162943.1 - 9 1310
BC070455.1 - 9 1316 30606785

View Full Product Details