Gene Transcripts

Gene Symbol
St6galnac1
Entrez Gene ID
20445
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Gene Aliases
Siat7a
Chromosome Location
Chr. 11: 116765025 - 116775507 on Build GRCm38
UniGene ID
Mm.436887
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011371.2 NP_035501.2 9 1576
GenBank mRNA AK155119.1 - 756
BC145374.1 - 9 1526 9053932
BE847090.1 - 3 240
Y11274.1 - 9 1576

View Full Product Details