Gene Transcripts

Gene Symbol
Six5
Entrez Gene ID
20475
Gene Name
sine oculis-related homeobox 5
Gene Aliases
Dmahp, MDMAHP, TrexBF
Chromosome Location
Chr. 7: 19094544 - 19098549 on Build GRCm38
UniGene ID
Mm.3410
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011383.1 NP_035513.1 2 1105
GenBank mRNA AK216841.1 - 1 44
D83146.1 - 2 856

View Full Product Details