Gene Transcripts

Gene Symbol
Slc13a2
Entrez Gene ID
20500
Gene Name
solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 2
Gene Aliases
Nadc1, mNaDC-1
Chromosome Location
Chr. 11: 78397276 - 78422280 on Build GRCm38
UniGene ID
Mm.274058
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022411.3 NP_071856.1 8 1180
GenBank mRNA AF201903.1 - 8 1180
AK140972.1 - 8 1231
BC013493.1 - 8 1205 4218337

View Full Product Details