Gene Transcripts

Gene Symbol
Sprr2e
Entrez Gene ID
20759
Gene Name
small proline-rich protein 2E
Gene Aliases
Chromosome Location
Chr. 3: 92352143 - 92353449 on Build GRCm38
UniGene ID
Mm.261596
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011471.2 NP_035601.1 2 543
GenBank mRNA AY158989.1 - 2 543

View Full Product Details