Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080965.1 | NP_001074434.1 | 4 | 404 | |
NM_001080966.1 | NP_001074435.1 | 4 | 690 | ||
NM_020572.2 | NP_065597.2 | 4 | 568 | ||
GenBank mRNA | AF054620.1 | - | 4 | 655 | |
BC064780.1 | - | 4 | 572 | 30476803 | |
BC100337.1 | - | 4 | 325 | ||
BG080448.2 | - | 4 | 381 | ||
DV648982.1 | - | 4 | 432 |