Gene Transcripts

Gene Symbol
Tcte2
Entrez Gene ID
21646
Gene Name
t-complex-associated testis expressed 2
Gene Aliases
AI429268, Tcte-2
Chromosome Location
Chr. 17: 13716436 - 13761394 on Build GRCm38
UniGene ID
Mm.486358
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022311.2 NP_071706.2 2 192
GenBank mRNA AK005879.1 - 177
AK008572.1 - 178
BC049646.1 - 2 184
BC092528.1 - 2 192 6741782
U46151.1 - 2 399

View Full Product Details