Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282006.1 | NP_001268935.1 | 3 | 407 | |
NM_001282007.1 | NP_001268936.1 | 3 | 356 | ||
NM_011569.3 | NP_035699.2 | 3 | 359 | ||
GenBank mRNA | AF081947.1 | - | 3 | 350 | |
AK005801.1 | - | 3 | 408 | ||
AK076858.1 | - | 3 | 337 | ||
BC050967.1 | - | 3 | 357 | ||
BC098181.1 | - | 3 | 317 | 6312556 |