Gene Transcripts

Gene Symbol
Tspan7
Entrez Gene ID
21912
Gene Name
tetraspanin 7
Gene Aliases
1200014P11Rik, A15, AI323365, Cd231, Mxs1, PE31, R74651, TALLA, Tm4sf2
Chromosome Location
Chr. X: 10485116 - 10596604 on Build GRCm38
UniGene ID
Mm.18590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019634.2 NP_062608.2 6 714
GenBank mRNA AF052492.1 - 6 662
AK004776.1 - 6 674
BC026516.1 - 5 513 4911317
BC050153.1 - 6 662
BC058603.1 - 6 670 6529011
D26483.1 - 6 662

View Full Product Details