Gene Transcripts

Gene Symbol
Vmn2r123
Entrez Gene ID
22300
Gene Name
vomeronasal 2, receptor 123
Gene Aliases
V2r1
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.377117
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009485.1 NP_033511.1 3 1025
GenBank mRNA AF011411.1 - 3 1025

View Full Product Details