Gene Transcripts
- Gene Symbol
- Vegfb
- Entrez Gene ID
- 22340
- Gene Name
- vascular endothelial growth factor B
- Gene Aliases
- VEGF-B, Vrf
- Chromosome Location
- Chr. 19: 6982472 - 6987651 on Build GRCm38
- UniGene ID
- Mm.15607
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001185164.1 |
NP_001172093.1
|
3, 4 |
547 |
|
NM_011697.3 |
NP_035827.1
|
3, 4 |
547 |
|
GenBank mRNA |
AK148188.1 |
- |
3, 4 |
546 |
|
AK178982.1 |
- |
1, 2 |
178 |
|
AK207652.1 |
- |
1, 2 |
174 |
|
AK208173.1 |
- |
1, 2 |
178 |
|
AK211743.1 |
- |
1, 2 |
179 |
|
AK212120.1 |
- |
1, 2 |
179 |
|
AK216634.1 |
- |
1, 2 |
178 |
|
BC046303.1 |
- |
3, 4 |
525 |
6492277 |
U43836.1 |
- |
3, 4 |
499 |
|
U43837.1 |
- |
3, 4 |
307 |
|
U48800.1 |
- |
3, 4 |
401 |
|
U52820.1 |
- |
3, 4 |
401 |
|
View Full Product Details