Gene Transcripts
- Gene Symbol
- Wbp2
- Entrez Gene ID
- 22378
- Gene Name
- WW domain binding protein 2
- Gene Aliases
- Chromosome Location
- Chr. 11: 116078573 - 116086997 on Build GRCm38
- UniGene ID
- Mm.284792
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016852.2 |
NP_058548.1
|
1, 2 |
106 |
|
GenBank mRNA |
AK004824.1 |
- |
1, 2 |
108 |
|
AK043037.1 |
- |
1, 2 |
80 |
|
AK079300.1 |
- |
1, 2 |
96 |
|
AK146572.1 |
- |
1, 2 |
108 |
|
AK150018.1 |
- |
1, 2 |
81 |
|
AK151368.1 |
- |
1, 2 |
98 |
|
AK162810.1 |
- |
1, 2 |
81 |
|
AK170317.1 |
- |
1, 2 |
107 |
|
AK170474.1 |
- |
1, 2 |
109 |
|
AK187444.1 |
- |
1, 2 |
35 |
|
BC031908.1 |
- |
1, 2 |
69 |
5365118 |
BC055058.1 |
- |
1, 2 |
126 |
6529458 |
U40826.1 |
- |
1, 2 |
106 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK044015.1 |
- |
|
96 |
|
Chr. - - [-] |
View Full Product Details