Gene Transcripts
- Gene Symbol
- Map3k19
- Entrez Gene ID
- 22625
- Gene Name
- mitogen-activated protein kinase kinase kinase 19
- Gene Aliases
- Ysk4
- Chromosome Location
- Chr. 1: 127814962 - 127855613 on Build GRCm38
- UniGene ID
- Mm.246566
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011737.1 |
NP_035867.1
|
7 |
2376 |
|
View Full Product Details