Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_194333.3 | NP_919314.2 | 8 | 1038 | |
GenBank mRNA | BC094893.1 | - | 8 | 1042 | |
BC117790.1 | - | 8 | 1025 | 40092152 | |
BC117791.1 | - | 8 | 1025 | 40092153 | |
U25739.1 | - | 8 | 1050 |