Gene Transcripts

Gene Symbol
Jtb
Entrez Gene ID
23922
Gene Name
jumping translocation breakpoint
Gene Aliases
Gm622
Chromosome Location
Chr. 3: 90231597 - 90235840 on Build GRCm38
UniGene ID
Mm.13912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206924.2 NP_996807.1 5 673
GenBank mRNA AB016490.1 - 5 618
AK004469.1 - 5 585
AK089048.1 - 5 585
AK146337.1 - 5 674
AK215601.1 - 3 187
BC008139.1 - 5 637 3582930
BG077727.2 - 4 239

View Full Product Details