Gene Transcripts

Gene Symbol
Lynx1
Entrez Gene ID
23936
Gene Name
Ly6/neurotoxin 1
Gene Aliases
AI838844, SLURP-2
Chromosome Location
Chr. 15: 74747856 - 74752979 on Build GRCm38
UniGene ID
Mm.257067
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011838.4 NP_035968.1 4 434
GenBank mRNA AF141377.1 - 4 408
AK013827.1 - 4 431
AK161907.1 - 4 464
AK162920.1 - 4 457
AK165295.1 - 4 438
AK165314.1 - 4 438
BC037541.1 - 4 481 5325123

View Full Product Details