Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004974.3 | NP_004965.1 | 3 | 1908 | |
XM_011541396.2 | XP_011539698.1 | 3 | 1902 | ||
XM_011541397.2 | XP_011539699.1 | 4 | 2183 | ||
XM_011541398.2 | XP_011539700.1 | 3 | 1950 | ||
XM_011541399.2 | XP_011539701.1 | 3 | 1991 | ||
XM_011541400.2 | XP_011539702.1 | 3 | 1921 | ||
XM_017001213.1 | XP_016856702.1 | 3 | 1883 | ||
GenBank mRNA | AK290061.1 | - | 3 | 1908 | |
L02752.1 | - | 1 | 1287 |