Gene Transcripts

Gene Symbol
Ciao1
Entrez Gene ID
26371
Gene Name
cytosolic iron-sulfur protein assembly 1
Gene Aliases
AW210570, Wdr39
Chromosome Location
Chr. 2: 127240938 - 127247816 on Build GRCm38
UniGene ID
Mm.45179
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025296.4 NP_079572.2 5 722
GenBank mRNA AK002314.1 - 5 667
AK004129.1 - 5 648
AK028376.1 - 5 649
BC004089.1 - 5 643 3593736

View Full Product Details