Gene Transcripts

Gene Symbol
Map3k1
Entrez Gene ID
26401
Gene Name
mitogen-activated protein kinase kinase kinase 1
Gene Aliases
MAPKKK1, MEKK1, Mekk
Chromosome Location
Chr. 13: 111746433 - 111818269 on Build GRCm38
UniGene ID
Mm.15918
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011945.2 NP_036075.2 4 999
GenBank mRNA AB014614.1 - 4 976
AF117340.1 - 4 999

View Full Product Details