Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039169.1 | NP_001034258.1 | 3 | 713 | |
NM_001039170.1 | NP_001034259.1 | 3 | 262 | ||
NM_023314.3 | NP_075803.2 | 3 | 262 | ||
GenBank mRNA | AF068116.1 | - | 3 | 176 | |
AK017221.1 | - | 3 | 714 | ||
AK018516.1 | - | 3 | 260 | ||
AK032965.1 | - | 3 | 266 | ||
AK138055.1 | - | 3 | 201 | ||
AK150495.1 | - | 3 | 201 | ||
AK199854.1 | - | 3 | 244 | ||
BC028903.1 | - | 3 | 312 | 4166275 | |
BC045153.1 | - | 3 | 196 | 6441327 | |
BC049077.1 | - | 3 | 230 | 5708556 | |
U01137.1 | - | 3 | 209 |