Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004982.3 | NP_004973.1 | 3 | 1502 | |
XM_005253358.4 | XP_005253415.1 | 4 | 1471 | ||
XM_017019283.1 | XP_016874772.1 | 4 | 1406 | ||
XM_017019284.1 | XP_016874773.1 | 4 | 1477 | ||
GenBank mRNA | AK291821.1 | - | 4 | 1310 | |
BC000544.2 | - | 3 | 1362 | 3161776 | |
D50312.1 | - | 3 | 1426 |