Gene Transcripts
- Gene Symbol
- Xlr5c
- Entrez Gene ID
- 27084
- Gene Name
- X-linked lymphocyte-regulated 5C
- Gene Aliases
- Xlr5
- Chromosome Location
- Chr. X: 73283361 - 73293970 on Build GRCm38
- UniGene ID
- Mm.255790
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031493.1 |
NP_113681.1
|
5, 6 |
426 |
|
GenBank mRNA |
BC119509.2 |
- |
5, 6 |
426 |
|
- Gene Symbol
- Xlr5a
- Entrez Gene ID
- 574438
- Gene Name
- X-linked lymphocyte-regulated 5A
- Gene Aliases
- Xlr5d
- Chromosome Location
- Chr. X: 73107621 - 73119524 on Build GRCm38
- UniGene ID
- Mm.435653
- Species
- Mus musculus
- Gene Symbol
- Xlr5b
- Entrez Gene ID
- 627081
- Gene Name
- X-linked lymphocyte-regulated 5B
- Gene Aliases
- OTTMUSG00000017646
- Chromosome Location
- Chr. X: 73146976 - 73158886 on Build GRCm38
- UniGene ID
- Mm.255790
- Species
- Mus musculus
View Full Product Details