Gene Transcripts

Gene Symbol
Pald1
Entrez Gene ID
27355
Gene Name
phosphatase domain containing, paladin 1
Gene Aliases
MMPAL, Pald, mKIAA1274
Chromosome Location
Chr. 10: 61319656 - 61383533 on Build GRCm38
UniGene ID
Mm.269699
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013753.2 NP_038781.2 18 2479
GenBank mRNA AK088577.1 - 18 2315
AK146489.1 - 18 2424
BC137695.1 - 17 2243 8860714
BC144846.1 - 17 2252 9053394
BC144847.1 - 17 2249 9053395
X99384.1 - 17 2264

View Full Product Details