Gene Transcripts

Gene Symbol
Igfbp7
Entrez Gene ID
29817
Gene Name
insulin-like growth factor binding protein 7
Gene Aliases
AGM, Fstl2, Mac25
Chromosome Location
Chr. 5: 77349240 - 77408045 on Build GRCm38
UniGene ID
Mm.233470
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159518.1 NP_001152990.1 6 1071
NM_008048.3 NP_032074.3 5 978
GenBank mRNA AB012886.1 - 5 979
AK002277.1 - 5 974
AK148749.1 - 5 975
AK156315.1 - 5 980
BC047202.1 - 5 890
BC092538.1 - 5 974 30284911
L75822.2 - 5 956

View Full Product Details