Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013895.4 | NP_038923.1 | 2, 3 | 408 | |
GenBank mRNA | AF144702.1 | - | 2, 3 | 410 | |
AK003054.1 | - | 2, 3 | 214 | ||
AK009041.1 | - | 2, 3 | 202 | ||
AK009152.1 | - | 2, 3 | 218 | ||
AK013230.1 | - | 2, 3 | 206 | ||
AK088984.1 | - | 2, 3 | 218 | ||
BC011436.1 | - | 2, 3 | 221 | 2812494 |