Gene Transcripts

Gene Symbol
Fbxw5
Entrez Gene ID
30839
Gene Name
F-box and WD-40 domain protein 5
Gene Aliases
AI159739, Fbw5
Chromosome Location
Chr. 2: 25500767 - 25505471 on Build GRCm38
UniGene ID
Mm.29170
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013908.4 NP_038936.1 2 261
GenBank mRNA AF176520.1 - 2 222
AK019715.2 - 2 262
AK154911.1 - 1 1200
BC010776.1 - 2 188 4211592

View Full Product Details