Gene Transcripts

Gene Symbol
Cbx8
Entrez Gene ID
30951
Gene Name
chromobox 8
Gene Aliases
Pc3
Chromosome Location
Chr. 11: 119038436 - 119040913 on Build GRCm38
UniGene ID
Mm.99953
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013926.1 NP_038954.1 5 1066
GenBank mRNA AB221673.1 - 5 1003
AF180370.1 - 5 1066
AK051905.1 - 496
BC014815.1 - 5 1016 4456898

View Full Product Details