Gene Transcripts
- Gene Symbol
- Siva1
- Entrez Gene ID
- 30954
- Gene Name
- SIVA1, apoptosis-inducing factor
- Gene Aliases
- CD27bp, Siva
- Chromosome Location
- Chr. 12: 112644828 - 112649152 on Build GRCm38
- UniGene ID
- Mm.289812
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013929.2 |
NP_038957.2
|
1, 2 |
253 |
|
GenBank mRNA |
AF033113.1 |
- |
1, 2 |
114 |
|
AF033114.1 |
- |
1, 2 |
201 |
|
AK010562.1 |
- |
1, 2 |
179 |
|
AK017570.1 |
- |
1, 2 |
214 |
|
AK153115.1 |
- |
1, 2 |
163 |
|
AK169200.1 |
- |
1, 2 |
206 |
|
AK184852.1 |
- |
1, 2 |
58 |
|
AK209128.1 |
- |
1, 2 |
58 |
|
BC012279.1 |
- |
1, 2 |
235 |
4013026 |
CB524656.1 |
- |
1, 2 |
402 |
|
View Full Product Details