Gene Transcripts

Gene Symbol
Spast
Entrez Gene ID
50850
Gene Name
spastin
Gene Aliases
Spg4, mKIAA1083
Chromosome Location
Chr. 17: 74337454 - 74391115 on Build GRCm38
UniGene ID
Mm.19804
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162870.1 NP_001156342.1 2 496
NM_016962.2 NP_058658.2 2 493
GenBank mRNA AJ246002.1 - 2 118
AK007793.1 - 2 273
AK044900.1 - 2 348
AK129282.1 - 2 470
BC046286.1 - 2 493 6441742

View Full Product Details